3-(1,3-Oxazol-5-yl)aniline

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-(5-Oxazolyl)aniline

OR29046-1G OR29046-250MG
76559-532EA 44 USD
76559-532 76562-178
3-(1,3-Oxazol-5-yl)aniline
3-(1,3-Oxazol-5-yl)aniline
Formula: C₉H₈N₂O
MW: 160.1700 g/mol
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00052191
CAS Number: 157837-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR