Ethyl-2-chloronicotinate 98%

Warning

Synonyms: Ethyl 2-chloropyridine-3-carboxylate

OR28967-50G OR28967-10G OR28967-250G
76572-794EA 82.4 USD
76572-794 76572-792 76572-790
Ethyl-2-chloronicotinate 98%
Ethyl 2-chloronicotinate
Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Boiling Pt: 78 °C/0.08mmHg
Density: 1.25 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00674129
CAS Number: 1452-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR