4-(1H-Pyrazol-5-yl)aniline

Supplier: APOLLO SCIENTIFIC

Warning

OR2888-250MG OR2888-1G OR2888-100MG
76572-734EA 31 USD
76572-734 76572-736 76572-738
4-(1H-Pyrazol-5-yl)aniline
4-(1H-Pyrazol-5-yl)aniline
Formula: C₉H₉N₃
Melting Pt: 110…115 °C
Storage Temperature: Ambient
MDL Number: MFCD09055175
CAS Number: 89260-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR