Formamidine acetate 99%

Supplier: APOLLO SCIENTIFIC

Warning

OR28877-500G OR28877-100G
76572-726EA 35 USD
76572-726 76572-728
Formamidine acetate 99%
Formamidine acetate
Formula: C₃H₈N₂O₂
Melting Pt: 157…161 °C
Density: 1.124 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012866
CAS Number: 3473-63-0
EINECS: 222-442-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR