(R)-(-)-Methyl-3-hydroxybutyrate

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Methyl (R)-3-hydroxybutyrate(R)-Methyl 3-hydroxybutanoate

OR28835-100G OR28835-10G OR28835-25G
76572-704EA 61 USD
76572-704 76572-706 76572-708
(R)-(-)-Methyl-3-hydroxybutyrate
(R)-(-)-Methyl 3-hydroxybutyrate
Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 72 °C/17mmHg
Density: 1.055 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00063289
CAS Number: 3976-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR