4,7-Dichloroquinoline

Supplier: APOLLO SCIENTIFIC

Warning

OR28817-25G OR28817-100G
76572-678EA 16 USD
76572-678 76572-680
4,7-Dichloroquinoline
4,7-Dichloroquinoline
Formula: C₉H₅Cl₂N
MW: 198.05 g/mol
Boiling Pt: 148 °C/10mmHg
Melting Pt: 81…87 °C
Flash Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00006774
CAS Number: 86-98-6
EINECS: 201-714-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR