2-Amino-5-methyl-1,3,4-thiadiazole

Supplier: APOLLO SCIENTIFIC
Warning

OR28803-5G
76572-672EA 13.33 USD
76572-672
2-Amino-5-methyl-1,3,4-thiadiazole
2-Amino-5-methyl-1,3,4-thiadiazole
Formula: C₃H₅N₃S
Melting Pt: 223…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00003110
CAS Number: 108-33-8
EINECS: 203-573-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR