1,3-Cyclohexanedione 99%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Cyclohexane-1,3-dione

OR28703-500G OR28703-1KG OR28703-100G
76572-604EA 25 USD
76572-604 76572-606 76572-608
1,3-Cyclohexanedione 99%
1,3-Cyclohexanedione
Formula: C₆H₈O₂
MW: 112.13 g/mol
Melting Pt: 101…105 °C
Density: 1.1 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001585
CAS Number: 504-02-9
EINECS: 207-980-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR