1,8-Dibromooctane 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: Octamethylendibromide

OR28631-100G
76561-330EA 81 USD
76561-330
1,8-Dibromooctane 98%
1,8-Dibromooctane
Formula: BrH₂C(CH₂)₆CH₂Br
MW: 272.02 g/mol
Boiling Pt: 270…272 °C
Melting Pt: 15…16 °C
Storage Temperature: Ambient
MDL Number: MFCD00000277
CAS Number: 4549-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR