2-Chloro-5-iodotoluene

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Chloro-4-iodo-2-methylbenzene

OR2862-1G OR2862-10G OR2862-5G
76572-522EA 33 USD
76572-522 76572-524 76572-520
2-Chloro-5-iodotoluene
2-Chloro-5-iodotoluene
Formula: C₇H₆ClI
MW: 252.48 g/mol
Boiling Pt: 239 °C
Density: 1.81 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03093903
CAS Number: 116632-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR