2-Chloro-3-nitropyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR28619-5G OR28619-100G OR28619-25G
76572-514EA 13.39 USD
76572-514 76572-516 76572-518
2-Chloro-3-nitropyridine
2-Chloro-3-nitropyridine
Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Melting Pt: 100…103 °C
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00006232
CAS Number: 5470-18-8
EINECS: 226-799-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR