Isoquinoline-4-amine

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-AminoisoquinolineIsoquinolin-4-amine

OR2818-5G OR2818-1G
76572-356EA 31 USD
76572-356 76572-358
Isoquinoline-4-amine
Isoquinoline-4-amine
Formula: C₉H₈N₂
MW: 144.1700 g/mol
Melting Pt: 107…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00034752
CAS Number: 23687-25-4
EINECS: 245-823-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR