1-Methyl-1H-pyrazolo[3,4-d]pyrimidin-4-ylamine

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-Amino-1-methyl-1H-pyrazolo[3,4-d]pyrimidine

OR2758-250MG OR2758-1G
76572-214EA 32 USD
76572-214 76572-216
1-Methyl-1H-pyrazolo[3,4-d]pyrimidin-4-ylamine
1-Methyl-1H-pyrazolo[3,4-d]pyrimidin-4-ylamine
Formula: C₆H₇N₅
MW: 149.16 g/mol
Melting Pt: 268…270 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00128302
CAS Number: 5334-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR