Methyl-3-bromoimidazo[1,2-a]pyridine-6-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 3-bromoimidazo[1,2-a]pyridine-6-carboxylate

OR2750-1G
76572-150EA 33 USD
76572-150
Methyl-3-bromoimidazo[1,2-a]pyridine-6-carboxylate
Methyl 3-bromoimidazo[1,2-a]pyridine-6-carboxylate
Formula: C₉H₇BrN₂O₂
MW: 255.0681 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07021501
CAS Number: 886361-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR