4'-(Benzyloxy)-2'-hydroxy-3'-methylacetophenone

Supplier: APOLLO SCIENTIFIC
Warning

OR27465-100MG
76572-122EA 51 USD
76572-122
4'-(Benzyloxy)-2'-hydroxy-3'-methylacetophenone
4'-(Benzyloxy)-2'-hydroxy-3'-methylacetophenone
Formula: C₁₆H₁₆O₃
Melting Pt: 89…91 °C
Storage Temperature: Ambient
CAS Number: 73640-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR