3,6-Dibromopyridazine

Supplier: APOLLO SCIENTIFIC

Warning

OR2744-5G OR2744-100G OR2744-25G
76572-094EA 18 USD
76572-094 76572-096 76572-098
3,6-Dibromopyridazine
3,6-Dibromopyridazine
Formula: C₄H₂Br₂N₂
MW: 237.88 g/mol
Melting Pt: 117…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00233947
CAS Number: 17973-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR