Methyl-5-bromonicotinate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Methyl 5-bromonicotinate

OR27415-100G OR27415-25G OR27415-5G
76572-070EA 13 USD
76572-070 76572-072 76572-074
Methyl-5-bromonicotinate
Methyl 5-bromonicotinate
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00173668
CAS Number: 29681-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR