Methyl-6-chloronicotinate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 6-chloronicotinate

OR27398-500G OR27398-100G OR27398-25G
76572-060EA 58 USD
76572-060 76572-058 76572-062
Methyl-6-chloronicotinate 98%
Methyl 6-chloronicotinate
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 83…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00023420
CAS Number: 73781-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR