5-(Methoxycarbonyl)-2-pyridine carboxylic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 5-(Methoxycarbonyl)picolinic acid

OR27346-1G
76572-032EA 13 USD
76572-032
5-(Methoxycarbonyl)-2-pyridine carboxylic acid
5-(Methoxycarbonyl)-2-pyridine carboxylic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00179343
CAS Number: 17874-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR