5-Methyl-2,1,3-benzothiadiazole

Supplier: APOLLO SCIENTIFIC
Warning

OR26809-250MG
76571-926EA 13 USD
76571-926
5-Methyl-2,1,3-benzothiadiazole
5-Methyl-2,1,3-benzothiadiazole
Formula: C₇H₆N₂S
Boiling Pt: 62…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00173945
CAS Number: 1457-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR