Sarcosine methyl ester hydrochloride 97+%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: H-Sar-OMe·HCl

OR26803-100G OR26803-25G OR26803-5G
76571-924EA 176 USD
76571-924 76571-922 76571-920
Sarcosine methyl ester hydrochloride 97+%
Sarcosine methyl ester hydrochloride
Formula: C₄H₉NO₂·HCl
MW: 139.58 g/mol
Melting Pt: 105…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00038876
CAS Number: 13515-93-0
EINECS: 236-853-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR