4'-Methoxy-3'-nitroacetophenone 98%

Supplier: APOLLO SCIENTIFIC

Warning

OR26752-10G OR26752-5G OR26752-1G
76571-902EA 15 USD
76571-902 76571-906 76571-904
4'-Methoxy-3'-nitroacetophenone 98%
4'-Methoxy-3'-nitroacetophenone
Formula: C₉H₉NO₄
Melting Pt: 97…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00017023
CAS Number: 6277-38-9
EINECS: 228-476-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR