4-Bromo-1,3-dimethyl-1H-pyrazole-5-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR25990-5G OR25990-1G
76571-802EA 123.6 USD
76571-802 76571-800
4-Bromo-1,3-dimethyl-1H-pyrazole-5-carboxylic acid
4-Bromo-1,3-dimethyl-1H-pyrazole-5-carboxylic acid
Formula: C₆H₇BrN₂O₂
MW: 219.0360 g/mol
Melting Pt: 230…232 °C
Storage Temperature: Ambient
MDL Number: MFCD02090873
CAS Number: 5775-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR