4-(Methylsulfonyl)benzoic acid 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-Methylsulfonylbenzoic acid

OR2590-100G OR2590-500G OR2590-25G
76571-736EA 360 USD
76571-736 76571-734 76571-732
4-(Methylsulfonyl)benzoic acid 98%
4-(Methylsulphonyl)benzoic acid
Formula: C₈H₈O₄S
MW: 200.21 g/mol
Melting Pt: 268…275 °C
Storage Temperature: Ambient
MDL Number: MFCD00007564
CAS Number: 4052-30-6
EINECS: 223-765-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR