5-Chloro-3-pyridineboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Chloropyridine-3-boronic acid(5-Chloropyridin-3-yl)boronic acid

OR2582-25G OR2582-5G OR2582-1G
76571-712EA 183 USD
76571-712 76571-710 76571-708
5-Chloro-3-pyridineboronic acid
5-Chloro-3-pyridineboronic acid
Formula: C₅H₅BClNO₂
MW: 157.36 g/mol
Melting Pt: 255…260 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06798243
CAS Number: 872041-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR