Diphenylphosphinic acid 99%

Supplier: APOLLO SCIENTIFIC

Warning

OR25790-25G OR25790-100G
76571-704EA 29 USD
76571-704 76571-706
Diphenylphosphinic acid 99%
Diphenylphosphinic acid
Formula: C₁₂H₁₁O₂P
MW: 218.19 g/mol
Melting Pt: 193…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00002132
CAS Number: 1707-03-5
EINECS: 216-948-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR