1,3-Cyclopentanedione

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Cyclopentane-1,3-dione

OR24952-25G OR24952-5G
76571-512EA 66 USD
76571-512 76571-510
1,3-Cyclopentanedione
1,3-Cyclopentanedione
Formula: C₅H₆O₂
MW: 98.1 g/mol
Melting Pt: 148…153 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001405
CAS Number: 3859-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR