2-Butylimidazole-4-carbaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR2486-250MG OR2486-1G
76571-480EA 13 USD
76571-480 76571-482
2-Butylimidazole-4-carbaldehyde
2-Butylimidazole-4-carbaldehyde
Formula: C₈H₁₂N₂O
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD01075030
CAS Number: 68282-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR