3-Bromocarbazole

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Bromo-9H-carbazole

OR24631-5G OR24631-25G
76571-442EA 32 USD
76571-442 76571-440
3-Bromocarbazole
3-Bromocarbazole
Formula: C₁₂H₈BrN
MW: 246.11 g/mol
Melting Pt: 196…201 °C
Storage Temperature: Ambient
MDL Number: MFCD00222621
CAS Number: 1592-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR