3-Coumaranone

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Benzofuran-3(2H)-one

OR24591-25G OR24591-5G OR24591-1G
76571-434EA 138 USD
76571-434 76571-432 76571-430
3-Coumaranone
3-Coumaranone
Formula: C₈H₆O₂
MW: 134.13 g/mol
Melting Pt: 97…103 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051810
CAS Number: 7169-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR