N-Acetylguanidine

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: N-Carbamimidoylacetamide1-Acetylguanidine

OR24582-25G OR24582-10G OR24582-5G
76571-428EA 149 USD
76571-428 76571-426 76571-424
N-Acetylguanidine
N-Acetylguanidine
Formula: C₃H₇N₃O
MW: 101.11 g/mol
Melting Pt: 181…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00075615
CAS Number: 5699-40-1
EINECS: 227-180-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR