6-Bromo-1,4-benzodioxane 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 6-Bromo-2,3-dihydro-1,4-benzodioxine6-Bromo-2,3-dihydrobenzo[b][1,4]dioxine

OR2430-5G OR2430-100G OR2430-25G
76571-328EA 18 USD
76571-328 76571-332 76571-330
6-Bromo-1,4-benzodioxane 98%
6-Bromo-1,4-benzodioxane
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 259…260 °C
Density: 1.598 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00040750
CAS Number: 52287-51-1
EINECS: 257-817-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR