Aluminon

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate)Aurin Tricarboxylic Acid Ammonium Salt

OR2429-250G OR2429-100G
76571-326EA 33 USD
76571-326 76571-324
Aluminon
Aluminon
Formula: C₂₂H₂₃N₃O₉
MW: 473.44 g/mol
Melting Pt: 220…225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00040925
CAS Number: 569-58-4
EINECS: 209-319-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR