2-(Methylsulfanyl)-1,4,5,6-tetrahydropyrimidinehydroiodide

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2-(Methylthio)-1,4,5,6-tetrahydropyrimidine hydroiodide

OR23426-5G OR23426-1G
76571-128EA 214 USD
76571-128 76571-126
2-(Methylsulfanyl)-1,4,5,6-tetrahydropyrimidinehydroiodide
2-(Methylsulfanyl)-1,4,5,6-tetrahydropyrimidinehydroiodide
Formula: C₅H₁₁IN₂S
MW: 258.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00829842
CAS Number: 5445-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR