1-Phenyl-1H-pyrazole-5-carboxylic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR23386-1G OR23386-250MG
76571-106EA 95 USD
76571-106 76571-104
1-Phenyl-1H-pyrazole-5-carboxylic acid 97%
1-Phenyl-1H-pyrazole-5-carboxylic acid
Formula: C₁₀H₈N₂O₂
Melting Pt: 182…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00463580
CAS Number: 1133-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR