Biphenyl-4,4'-dicarboxylic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: [1,1'-Biphenyl]-4,4'-dicarboxylic acid4,4'-Biphenyldicarboxylic acid

OR2318-5G OR2318-25G
76570-974EA 13 USD
76570-974 76570-976
Biphenyl-4,4'-dicarboxylic acid
Biphenyl-4,4'-dicarboxylic acid
Formula: C₁₄H₁₀O₄
MW: 242.23 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00002554
CAS Number: 787-70-2
EINECS: 212-328-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR