6-Bromoquinoxaline

Supplier: APOLLO SCIENTIFIC

Danger

OR23162-1G OR23162-500MG OR23162-5G
76570-954EA 37 USD
76570-954 76570-956 76570-958
6-Bromoquinoxaline
6-Bromoquinoxaline
Formula: C₈H₅BrN₂
MW: 209.0427 g/mol
Melting Pt: 48…53 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00837757
CAS Number: 50998-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR