5-Phenyl-2-thiopheneboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (5-Phenylthiophen-2-yl)boronic acid

OR23152-1G
76570-940EA 33 USD
76570-940
5-Phenyl-2-thiopheneboronic acid
5-Phenyl-2-thiopheneboronic acid
Formula: C₁₀H₉BO₂S
MW: 204.0500 g/mol
Melting Pt: 140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01114643
CAS Number: 306934-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR