2,1,3-Benzothiadiazole-5-carbaldehyde

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Benzo[c][1,2,5]thiadiazole-5-carbaldehyde

OR23127-5G OR23127-1G OR23127-10G
76570-922EA 140 USD
76570-922 76570-924 76570-926
2,1,3-Benzothiadiazole-5-carbaldehyde
2,1,3-Benzothiadiazole-5-carbaldehyde
Formula: C₇H₄N₂OS
MW: 164.1800 g/mol
Melting Pt: 89…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00457949
CAS Number: 71605-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR