6-Quinolinecarboxylic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR23075-1G OR23075-10G OR23075-25G
76570-880EA 87 USD
76570-880 76570-876 76570-878
6-Quinolinecarboxylic acid 98%
6-Quinolinecarboxylic acid
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Melting Pt: 293…296 °C
Storage Temperature: Ambient
MDL Number: MFCD00047613
CAS Number: 10349-57-2
EINECS: 233-761-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR