1-Cyclohexenylboronic acid pinacol ester 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(Cyclohex-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane1-Cyclohexene-1-boronic acid pinacol ester

OR2302-5G OR2302-1G
76570-836EA 28 USD
76570-836 76570-838
1-Cyclohexenylboronic acid pinacol ester 95%
1-Cyclohexenylboronic acid pinacol ester
Formula: C₁₂H₂₁BO₂
MW: 208.1049 g/mol
Boiling Pt: 125 °C/0.23mmHg
Density: 0.968 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD05663845
CAS Number: 141091-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR