4-Bromo-3,5-dimethyl-1H-pyrazole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Bromo-3,5-dimethylpyrazole

OR23001-25G OR23001-5G OR23001-100G
76570-812EA 14.35 USD
76570-812 76570-814 76570-816
4-Bromo-3,5-dimethyl-1H-pyrazole
4-Bromo-3,5-dimethyl-1H-pyrazole
Formula: C₅H₇BrN₂
MW: 175.03 g/mol
Melting Pt: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00005242
CAS Number: 3398-16-1
EINECS: 222-263-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR