4-Amino-N,N-dimethylbenzenesulfonamide

Supplier: APOLLO SCIENTIFIC
Warning

OR2276-1G OR2276-5G OR2276-250MG
76570-730EA 17 USD
76570-730 76570-732 76570-734
4-Amino-N,N-dimethylbenzenesulfonamide
4-Amino-N,N-dimethylbenzenesulphonamide
Formula: C₈H₁₂N₂O₂S
MW: 200.26 g/mol
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00031428
CAS Number: 1709-59-7
EINECS: 216-970-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR