1-Bromoadamantane 98%
Supplier: APOLLO SCIENTIFIC
Synonyms:
1-Adamantyl bromide
OR22505-100G
OR22505-25G
76570-660EA
16
USD
76570-660
76570-662
1-Bromoadamantane 98%
1-Bromoadamantane
Formula:
C₁₀H₁₅Br MW: 215.13 g/mol Melting Pt: 116…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074721 CAS Number: 768-90-1 EINECS: 212-200-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...