2'-Hydroxy-4',6'-dimethoxyacetophenone 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(2-Hydroxy-4,6-dimethoxyphenyl)ethan-1-one

OR22374-5G OR22374-25G OR22374-50G
76570-630EA 50 USD
76570-630 76570-632 76570-634
2'-Hydroxy-4',6'-dimethoxyacetophenone 99%
2'-Hydroxy-4',6'-dimethoxyacetophenone
Formula: C₁₀H₁₂O₄
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00017243
CAS Number: 90-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR