4-Chloro-2,6-diaminopyrimidine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 6-Chloropyrimidine-2,4-diamine2,4-Diamino-6-chloropyrimidine

OR22082-25G
76570-514EA 13 USD
76570-514
4-Chloro-2,6-diaminopyrimidine
4-Chloro-2,6-diaminopyrimidine
Formula: C₄H₅ClN₄
MW: 144.56 g/mol
Melting Pt: 199…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00006097
CAS Number: 156-83-2
EINECS: 205-863-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR