4-Hexylaniline

Supplier: APOLLO SCIENTIFIC

Warning

OR22069-1G OR22069-25G OR22069-5G
76560-848EA 22 USD
76560-848 76561-340 76560-498
4-Hexylaniline
4-Hexylaniline
Formula: C₁₂H₁₉N
MW: 177.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007927
CAS Number: 33228-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR