2,6-Di-tert-butylpyridine

Supplier: APOLLO SCIENTIFIC

Warning

OR21937-1G
76570-448EA 14 USD
76570-448
2,6-Di-tert-butylpyridine
2,6-Di-tert-butylpyridine
Formula: C₁₃H₂₁N
MW: 191.32 g/mol
Boiling Pt: 100…101 °C/23mmHg
Melting Pt: 44288 °C
Density: 0.852 g/cm³ (25 °C)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00006306
CAS Number: 585-48-8
EINECS: 209-557-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR