4-Hydroxy-3-nitropyridine-N-oxide 97+%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-Hydroxy-3-nitropyridine 1-oxide

OR2037-5G OR2037-1G
76570-054EA 30 USD
76570-054 76570-056
4-Hydroxy-3-nitropyridine-N-oxide 97+%
4-Hydroxy-3-nitropyridine N-oxide
Formula: C₅H₄N₂O₄
MW: 156.0963 g/mol
Melting Pt: 225…226 °C
Storage Temperature: Ambient
MDL Number: MFCD00234062
CAS Number: 31872-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR