Pyridine-2-carboxaldoxime

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2-Pyridinecarbaldehyde oxime

OR2013-5G
76570-022EA 43 USD
76570-022
Pyridine-2-carboxaldoxime
Pyridine-2-carboxaldoxime
Formula: C₆H₆N₂O
MW: 122.13 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00006354
CAS Number: 873-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR